Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2-fluoro-3-nitrobenzene, 97%

Catalog No. AAH6198914
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6198914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6198914 Supplier Thermo Scientific Chemicals Supplier No. H6198914
Only null left
Add to Cart

CAS: 2106-49-2 | C6H3ClFNO2 | 175.54 g/mol

1-Chloro-2-fluoro-3-nitrobenzene is used as pharmaceutical intermediates, agrochemical intermediates, intermediates of medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-fluoro-3-nitrobenzene is used as pharmaceutical intermediates, agrochemical intermediates, intermediates of medicine.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing a
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2106-49-2
Molecular Formula C6H3ClFNO2
Molecular Weight (g/mol) 175.54
MDL Number MFCD00069417
InChI Key RBAHXNSORRGCQA-UHFFFAOYSA-N
Synonym 3-chloro-2-fluoronitrobenzene, 2-fluoro-3-chloronitrobenzene, 1-chloro-2-fluoro-3-nitro-benzene, 2-fluoro-3-chloro-nitrobenzene, benzene, chlorofluoronitro, ccris 9050, 3-chloro-2-fluoro-1-nitrobenzene, pubchem2483, chlorofluoronitrobenzene, acmc-1bn8r
PubChem CID 75016
IUPAC Name 1-chloro-2-fluoro-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(Cl)=C1F

Specifications

Color Yellow
Quantity 25 g
UN Number UN2810
Solubility Information Soluble in water.
Formula Weight 175.54
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-fluoro-3-nitrobenzene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 218-285-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.