Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Cyclohexene-1-carboxylic acid, 97%
Click to view available options
Quantity:
1 g
5 g
25 g
Description
1-Cyclohexene-1-carboxylic acid is an intermediate in benzoic acid anaerobic decomposition by methanogenic consortium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Cyclohexene-1-carboxylic acid is an intermediate in benzoic acid anaerobic decomposition by methanogenic consortium.
Solubility
Soluble in water (0.7 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
1-Cyclohexene-1-carboxylic acid is an intermediate in benzoic acid anaerobic decomposition by methanogenic consortium.
Solubility
Soluble in water (0.7 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Specifications
Specifications
| Melting Point | 30°C to 34°C |
| Density | 1.1 |
| Boiling Point | 138°C to 140°C (14 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.499 |
| Quantity | 1 g |
| UN Number | UN3261 |
| Beilstein | 1905520 |
| Solubility Information | Soluble in water (0.7g/L at 20°C). |
| Formula Weight | 126.16 |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction