Learn More
1-Decyne, 98%
Description
1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.
Solubility
Not miscible in water.
Notes
Incompatible with strong oxidizing agents. Store at 0-6°C.
Specifications
Specifications
| Melting Point | -44°C |
| Density | 0.767 |
| Boiling Point | 174°C to 176°C |
| Flash Point | 50°C (122°F) |
| Odor | Petrol-like |
| Refractive Index | 1.427 |
| Quantity | 5 g |
| UN Number | UN3295 |
| Beilstein | 1236372 |
| Solubility Information | Not miscible in water. |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H318 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S. |
| EINECSNumber | 212-132-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.