Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Deoxy-1-nitro-D-iditol hemihydrate, 99%, Thermo Scientific™

Catalog No. AC353831000
Click to view available options
Quantity:
100 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207121-57-1
Molecular Formula C6H15NO8
Molecular Weight (g/mol) 229.185
MDL Number MFCD00191971
InChI Key OHFQXMNDKUHFDU-QAYODLCCSA-N
Synonym 1-deoxy-1-nitro-d-iditol hemihydrate, 2r,3s,4s,5r-6-nitrohexane-1,2,3,4,5-pentol hydrate
PubChem CID 73995059
IUPAC Name (2R,3S,4S,5R)-6-nitrohexane-1,2,3,4,5-pentol;hydrate
SMILES C(C(C(C(C(CO)O)O)O)O)[N+](=O)[O-].O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 100 mg
Specific Rotation +2.5° to +4.5° (20°C, 589nm) (c=4, H2O)
Formula Weight 220.18
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 1-Deoxy-1-nitro-D-iditol hemihydrate, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.