Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Deoxy-1-nitro-L-iditol hemihydrate, 99%, Thermo Scientific™

Catalog No. AC353721000
Click to view available options
Quantity:
100 mg

Chemical Identifiers

CAS 207226-23-1
Molecular Formula C12H28N2O15
Molecular Weight (g/mol) 440.36
MDL Number MFCD00191972
InChI Key HQAUHMYCAWVBKQ-UHFFFAOYNA-N
Synonym 1-deoxy-1-nitro-l-iditol hemihydrate, 2s,3r,4r,5s-6-nitrohexane-1,2,3,4,5-pentol hydrate
PubChem CID 73995060
IUPAC Name (2S,3R,4R,5S)-6-nitrohexane-1,2,3,4,5-pentol;hydrate
SMILES O.OCC(O)C(O)C(O)C(O)C[N+]([O-])=O.OCC(O)C(O)C(O)C(O)C[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 89°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Specific Rotation -4.5
Formula Weight 220.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1-Deoxy-1-nitro-L-iditol hemihydrate, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.