Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylimidazolium diethyl phosphate, 98%

Catalog No. aah2740006
Click to view available options
Quantity:
5 g
25 g

CAS: 848641-69-0 | C10H21N2O4P | 264.26 g/mol

It finds its application in improving enzymatic hydrolysis of wheat straw and in the enzymatic in situ saccharification of cellulose in aqueous-ionic liquid media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in improving enzymatic hydrolysis of wheat straw and in the enzymatic in situ saccharification of cellulose in aqueous-ionic liquid media.

Solubility
Fully miscible in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 848641-69-0
Molecular Formula C10H21N2O4P
Molecular Weight (g/mol) 264.26
MDL Number MFCD09953486
InChI Key HQWOEDCLDNFWEV-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium diethyl phosphate, 1-ethyl-3-methylimidazolium diethylphosphate, dsstox_cid_27865, dsstox_rid_82618, dsstox_gsid_47889, emim dep, basionics™ lq 11, 1-ethyl-3-methylimidazolium-diethyl phosphate, 1-ethyl-3-methylimidazolium diethyl phosphate hplc
PubChem CID 53316406
IUPAC Name diethyl phosphate;1-ethyl-3-methylimidazol-3-ium
SMILES CCN1C=C[N+](C)=C1.CCOP([O-])(=O)OCC

Specifications

Melting Point 19°C to 21°C
Density 1.157
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Fully miscible in water.
Formula Weight 264.26
Percent Purity 98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium diethyl phosphate
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.