Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Ethyl-3-methylpyridinium Ethyl Sulfate 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
| Quantity | 25 g |
| Formula Weight | 247.31 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Ethyl-3-methylpyridinium Ethyl Sulfate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction