Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heptanol, 99%

Catalog No. p-7020735
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 111-70-6 | C7H16O | 116.20 g/mol

1-Heptanol, is used in the fragrance industry due to its pleasant smell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptanol, is used in the fragrance industry due to its pleasant smell.

Solubility
Soluble in water 1.7 g/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-70-6
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00002986
InChI Key BBMCTIGTTCKYKF-UHFFFAOYSA-N
Synonym 1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
PubChem CID 8129
IUPAC Name heptan-1-ol
SMILES CCCCCCCO

Specifications

Melting Point -34°C
Density 0.82
Boiling Point 175°C to 176°C
Flash Point 73°C (163°F)
Odor Characteristic
Assay Percent Range 99%
Refractive Index 1.424
Linear Formula CH3(CH2)6OH
Quantity 100 mL
Beilstein 1731686
Merck Index 14,4661
Solubility Information Soluble in water 1.7g/L at 25°C.
Formula Weight 116.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Heptanol
Show More Show Less
Hazard Category H227-H319
Hazard Statement GHS H Statement
H311-H227-H302-H332-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
EINECSNumber 203-897-9
RTECSNumber MK0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.