Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Hexanesulfonic Acid Sodium Salt, HPLC Grade, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
25 g
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Description
- For Ion-Pair Chromatography of Basic Compounds
Specifications
Specifications
Packaging | Wide Mouth Amber Glass Bottle |
Quantity | 500 g |
Specific Gravity | 1L = 0.00kg |
IUPAC Name | sodium hexane-1-sulfonate |
Formula Weight | 188.22 |
Grade | HPLC |
Chemical Name or Material | 1-Hexanesulfonic Acid Sodium Salt |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction