Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-hexanesulfonic acid, sodium salt hydrate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
2832-45-3
C6H13NaO3S
188.22
MFCD00007542
QWSZRRAAFHGKCH-UHFFFAOYSA-M
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
23677630
sodium;hexane-1-sulfonate
[Na+].CCCCCCS([O-])(=O)=O
2832-45-3
C6H13NaO3S
188.22
MFCD00007542
QWSZRRAAFHGKCH-UHFFFAOYSA-M
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
23677630
sodium;hexane-1-sulfonate
[Na+].CCCCCCS([O-])(=O)=O
Vapor Pressure Negligible
Color White
Loss on Drying 9.6% max. (100°C, 3 hrs) (vacuum)
Absorbance 5.5 w/v% aq. soln., pH 3, H3PO4, 0.06 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm, 5.5 w/v% aq. soln., pH 3, H3PO4 at 0nm
Density Negligible
Infrared Spectrum Authentic
Melting Point >300.0°C
Packaging Glass bottle
Quantity 100g
Assay Percent Range 99%
Linear Formula CH3(CH2)5SO3Na·xH2O
Solubility Information (10% in water) Clear colorless
Formula Weight 188.22
Physical Form Crystalline Flakes or Solid
Percent Purity 99%
Chemical Name or Material 1-Hexanesulfonic acid, sodium salt hydrate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning