Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodooctane, 98+%, stab. with copper

Catalog No. aab2221414
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 629-27-6 | C8H17I | 240.128 g/mol

Alkylation of [2,2?-([2,2?-bipyridine]-6,6?-diyl)bis[phenolato]-N,N?,O,O?]nickel(II)was performed using the catalytic reduction of 1-iodooctane. Primary alkyl iodide 1-iodooctane reacts with lithium aluminum deuteride (LAD) in a 1:0.2 ratio to form 1-deuteriooctane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alkylation of [2,2′-([2,2′-bipyridine]-6,6′-diyl)bis[phenolato]-N,N′,O,O′]nickel(II)was performed using the catalytic reduction of 1-iodooctane. Primary alkyl iodide 1-iodooctane reacts with lithium aluminum deuteride (LAD) in a 1:0.2 ratio to form 1-deuteriooctane.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-27-6
Molecular Formula C8H17I
Molecular Weight (g/mol) 240.128
MDL Number MFCD00001105
InChI Key UWLHSHAHTBJTBA-UHFFFAOYSA-N
Synonym n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctane
PubChem CID 12380
IUPAC Name 1-iodooctane
SMILES CCCCCCCCI

Specifications

Melting Point -46°C
Density 1.325
Boiling Point 225°C to 226°C
Flash Point 95°C (203°F)
Refractive Index 1.4878
Linear Formula CH3(CH2)7I
Quantity 25 g
Beilstein 1697479
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 240.13
Percent Purity ≥98%
Chemical Name or Material 1-Iodooctane, stabilized with copper
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 211-084-5
RTECSNumber RG9700100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.