Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methoxy-2-propanol Acetate, 97%, Spectrum™ Chemical
SDP

Catalog No. 18600055
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can

M2634, 108-65-6, C6H12O3

Spectrum™ Chemical 1-Methoxy-2-propanol Acetate

Chemical Identifiers

CAS 108-65-6
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
InChI Key LLHKCFNBLRBOGN-UHFFFAOYNA-N
IUPAC Name 1-methoxypropan-2-yl acetate
SMILES COCC(C)OC(C)=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 4 L
UN Number UN3272
Percent Purity 97%
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.