Alfa Aesar™ 1-Methylsulfonyl-1H-benzotriazole, 97%

Manufacturer: Alfa Aesar™ H5338603

Quantity 1g

 View more versions of this product

Catalog No. AAH5338603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 37073-15-7
    Molecular Formula C7H7N3O2S
    Formula Weight 197.21
    MDL Number MFCD00595638
    Chemical Name or Material 1-Methylsulfonyl-1H-benzotriazole

    Specifications

    Formula Weight 197.21
    Melting Point 106° to 108°C
    Molecular Formula C7H7N3O2S
    MDL Number MFCD00595638
    CAS 37073-15-7
    Health Hazard 1 Danger
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H301-H318
    Toxic if swallowed.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P310-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 1-Methylsulfonyl-1H-benzotriazole
    Assay 97%

    Description

    Applications
    1-Methylsulfonyl-1H-benzotriazole is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    GHS H Statement
    H301-H318
    Toxic if swallowed.
    Causes serious eye damage.

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P310-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit