Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Methylsulfonyl)piperazine, 97%

Catalog No. AAH6387014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6387014 25 g
AAH6387006 5 g
AAH6387022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6387014 Supplier Thermo Scientific Chemicals Supplier No. H6387014
Add to Cart

CAS: 55276-43-2 | C5H13N2O2S | 165.23 g/mol

1-(Methylsulfonyl)piperazine is employed as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Methylsulfonyl)piperazine is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55276-43-2
Molecular Formula C5H13N2O2S
Molecular Weight (g/mol) 165.23
MDL Number MFCD02153225
InChI Key ZZAKLGGGMWORRT-UHFFFAOYSA-O
Synonym 1-methylsulfonyl piperazine, 1-methanesulfonylpiperazine, 1-methanesulfonyl-piperazine, 1-methylsulfonyl-piperazine, 1-methylsulphonyl piperazine, methylsulfonyl piperazine, piperazine, 1-methylsulfonyl, 1-n-methanesulfonylpiperazine, 4-methylsulfonylpiperazine, n-mesylpiperazine
PubChem CID 709161
IUPAC Name 1-methylsulfonylpiperazine
SMILES CS(=O)(=O)N1CC[NH2+]CC1

Specifications

Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 164.23
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(Methylsulfonyl)piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.