Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthyl trifluoromethanesulfonate, 97%

Catalog No. p-7052634
Click to view available options
Quantity:
1 g
5 g

CAS: 99747-74-7 | C11H7F3O3S | 276.229 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99747-74-7
Molecular Formula C11H7F3O3S
Molecular Weight (g/mol) 276.229
MDL Number MFCD00192340
InChI Key WQWUQDVFRYMMCY-UHFFFAOYSA-N
Synonym 1-naphthyl trifluoromethanesulfonate, 1-naphthyl triflate, 1-naphthyl-triflate, 1-naphthalenyl triflate, acmc-20aot0, 1-trifluoromethanesulfonyl naphthalene
PubChem CID 378884
IUPAC Name naphthalen-1-yl trifluoromethanesulfonate
SMILES C1=CC=C2C(=C1)C=CC=C2OS(=O)(=O)C(F)(F)F

Specifications

Color Red
Density 1.405 g/mL
Boiling Point 97°C to 98°C (0.3 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.52
Quantity 1 g
UN Number UN3265
Formula Weight 276.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Naphthyl trifluoromethanesulfonate
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.