Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octanesulfonic acid, sodium salt, HPLC grade

Catalog No. p-3211410
Click to view available options
Quantity:
5 g
25 g
100 g
500 g

1-Octanesulfonic acid, sodium salt, HPLC grade, Quantity: 25g, White, Melting Point: >300.0 deg.C, Molecular Weight: 216.28, Percent Purity: >-98.0% | CAS: 5324-84-5 | C8H17NaO3S | 216.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.27
MDL Number MFCD00007544
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
PubChem CID 23669624
SMILES [Na+].CCCCCCCCS([O-])(=O)=O

Specifications

Absorbance (0.25M aqueous solution), (0.25 M aqueous solution) at 0nm, 0.04 max. at 260nm, 0.05 max. at 240nm, 0.05 max. at 250nm, 0.075 max. at 230nm, 0.1 max. at 220nm, 0.2 max. at 210nm, 0.3 max. at 200nm
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3(CH2)7SO3Na
Quantity 5 g
Solubility Information Solubility in water: freely soluble. Other solubilities: 4g/L in methanol
IUPAC Name sodium;octane-1-sulfonate
Formula Weight 216.28
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material 1-Octanesulfonic acid, sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-195-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.