Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanesulfonic acid sodium salt, monohydrate; 99.9%, For HPLC analysis, MP Biomedicals™

Catalog No. p-4404852 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. ICN15181325

Supplier: MP Biomedicals, Inc 0215181325

Only null left
Add to Cart
Add to Cart

Specifications

Item Name 1-Pentanesulfonic Acid, Sodium Salt, Monohydrate
Content And Storage Store at Room Temperature.
Form Crystalline Powder
Quantity 25 g
Appearance White
Molecular Formula C5H11NaO3S
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
SMILES [Na+].CCCCCS([O-])(=O)=O
IUPAC Name sodium pentane-1-sulfonate
Molecular Weight (g/mol) 174.19
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.