Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanesulfonic Acid Sodium Salt, Monohydrate, HPLC Grade, 98%, Spectrum™ Chemical
SDP

Catalog No. 18600074 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail

P1043, 207605-40-1, C5H11O3SNa·H2O

Spectrum™ Chemical 1-Pentanesulfonic Acid Sodium Salt, Monohydrate, HPLC Grade

Chemical Identifiers

CAS 207605-40-1
Molecular Formula C5H11O3S
Molecular Weight (g/mol) 151.20
InChI Key RJQRCOMHVBLQIH-UHFFFAOYSA-M
IUPAC Name pentane-1-sulfonate
SMILES CCCCCS([O-])(=O)=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98%
Grade HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.