Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-Butoxycarbonyl)-2-piperidone 95.0+%, TCI America™
SDP

Catalog No. B37885G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 85908-96-9
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD02179046
InChI Key ULMHMJAEGZPQRY-UHFFFAOYSA-N
Synonym 1-boc-2-piperidone, n-boc-2-piperidone, 1-boc-1-azacyclohexan-2-one, 1-tert-butoxycarbonyl-2-piperidone, n-tert-butoxycarbonyl-2-piperidone, tert-butyl 2-oxo-1-piperidinecarboxylate, 1-piperidinecarboxylic acid, 2-oxo-, 1,1-dimethylethyl ester, tert-butyl 2-oxopiperidinecarboxylate, boc-piperidinone, n-boc piperidone
PubChem CID 7577838
ChEBI CHEBI:59556
IUPAC Name tert-butyl 2-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCCC1=O

Specifications

Boiling Point 110°C
Quantity 5 g
Formula Weight 199.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-(tert-Butoxycarbonyl)-2-piperidone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.