Learn More
United Scientific 10 mL Polypropylene Measuring Cylinder with Round Base
Shop All United Scientific Supplies, Inc Products
Click to view available options
Capacity (Metric):
10 mL
Description
- Polypropylene one-piece measuring cylinders have raised graduations for easy readability
- Autoclavable and have excellent contact clarity
- Broad round base provides good stability
Specifications
Specifications
Capacity (Metric) | 10 mL |
Material | Polypropylene |
Base Type | Round |