Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 100% Graphite Ferrules

Catalog No. p-3336780
Click to view available options
For Use With (Equipment):
For Thermo M4 Nut
Description:
0.25mm I.D.
0.32mm I.D.
0.53mm I.D.

Composed of a soft material that is porous to oxygen, making them suitable for most applications except GC/MS interface connections

  • Easy-to-use
  • Form a soft grip with the column and provide a stable seal

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) For Thermo M4 Nut
Description 0.25mm I.D.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.