Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEUROD1 Monoclonal Antibody (3E10F1), CoraLite594, Proteintech
SDP

Catalog No. 502333376 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-3376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-3376 Supplier Proteintech Group Inc Supplier No. CL59466691100UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NEUROD1 Monoclonal antibody specifically detects NEUROD1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry

Specifications

Antigen NEUROD1
Applications Immunocytochemistry
Classification Monoclonal
Clone 3E10F1
Concentration 1.2 mg/mL
Conjugate CoraLite 594
Formulation PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Accession No. Q13562
Gene Alias BETA2; BHF-1; BHLHA3; MODY6; NEUROD; NEUROD1
Host Species Mouse
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4760
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.