Learn More
Invitrogen™ PKC eta Recombinant Rabbit Monoclonal Antibody (JE32-41)

Description
Positive Control: MCF7 cell lysate, A431 cell lysate, COS-1 cell lysate, mouse brain tissue lysate, rat brain tissue lysate. Tissue Specificity: Low tissue specificity. Subcellular Location: Cytoplasm. Sequence Similarities: 97% Mouse/Rat. Predicted band size: 78 kDa.
Specifications
Specifications
| Antigen | PKC eta |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | JE32-41 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | PRKCH |
| Gene Accession No. | P23298, P24723, Q64617 |
| Gene Alias | LOW QUALITY PROTEIN: protein kinase C eta type; MGC26269; MGC5363; nPKC-eta; PKC; Pkch; PKCL; PKC-L; PRKCH; PRKCL; protein kinase C eta; protein kinase C eta type; protein kinase C eta type pseudogene; protein kinase C theta; protein kinase C, eta; protein kinase C-eta; Unknown (protein for MGC:140784) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.