Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ 10nm Colloidal Gold Nanoparticles 10OD
SDP

Catalog No. p-7111152 Shop All Novus Biologicals Products
Click to view available options
Quantity:
20 mL
100 mL

10nm colloidal gold nanoparticles ultra high quality, with narrow size distribution and uniform shape

  • 10nm colloidal gold nanoparticles are ultra high quality, with narrow size distribution and uniform shape, supplied in range of sizes and concentrations for maximum flexibility

Specifications

Content And Storage Store at 4°C. Do not freeze.
Quantity 20 mL
Kit Contents One 20mL bottle at 10 OD
Product Type 10nm Colloidal Gold Nanoparticles 10OD Antibody Labeling Kit

For research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.