Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Assay Buffer (10X)

Catalog No. T2187
Encompass_Preferred
Click to view available options
Quantity:
150 mL

Used in immunoassays and in Western and Southern blotting for alkaline phosphatase Tropix™ substrate conjugates.

  • Quantity: 150mL
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Buffer
Quantity 150 mL
Form Liquid
Substrate Type Tropix
Content And Storage Store the product at 4°C.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.