Learn More
Invitrogen™ BlueJuice™ Gel Loading Buffer (10X)

Description
Includes
3 vials BlueJuice™ Gel Loading Buffer (10X), 1mL each
- Designed for easy loading and tracking of DNA samples in agarose or native polyacrylamide gels
- Recommended concentration for use with DNA samples run on agarose gels is 2X (one part buffer plus four parts sample)R
- recommended concentration is 1X for acrylamide gels
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | • 3 x 1 mL 10X BlueJuice Gel Loading Buffer Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C. |
| Buffer | Sample Loading Buffers |
| Concentration | 10X |
| Gel Type | Agarose |
| Label or Dye | Bromophenol Blue |
| Product Type | Gel Loading Buffer |
| Sample Loading Volume | 1 mL |
| Gel Compatibility | Agarose Gels |
| Product Line | BlueJuice |
| Quantity | 3 x 1 mL |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.