Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11 beta-HSD1 Antibody, Novus Biologicals™
SDP

Catalog No. NB300934 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300934 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300934 Supplier Novus Biologicals Supplier No. NB300934
Add to Cart
Add to Cart

Goat Polyclonal Antibody

11 beta-HSD1 Polyclonal specifically detects 11 beta-HSD1 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen 11 beta-HSD1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_005516
Gene Alias 11-beta-hydroxysteroid dehydrogenase 1, EC 1.1.1, EC 1.1.1.146, HDL, HSD11HSD11B, HSD11LMGC13539, hydroxysteroid (11-beta) dehydrogenase 1, member 1, SDR26C1
Gene Symbols HSD11B1
Host Species Goat
Immunogen Peptide with sequence CTSYNMDRFINK corresponding to C-Terminus according to NP_005516.1, NP_861420.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3290
Test Specificity This antibody is expected to recognize human HSD11B1 protein. Both NP_005516.1 and NP_861420.1 variants encode the same protein.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.