Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
11-Bromo-1-undecene, 90+%
Click to view available options
Quantity:
5 g
25 g
Description
11-Bromo-1-undecene is used in advanced polymer dielectrics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
11-Bromo-1-undecene is used in advanced polymer dielectrics.
Solubility
Soluble in water, 0.2422 mg/L @ 25°C (est).
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
11-Bromo-1-undecene is used in advanced polymer dielectrics.
Solubility
Soluble in water, 0.2422 mg/L @ 25°C (est).
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications
Specifications
| Density | 1.063 |
| Boiling Point | 149°C to 150°C (35 mm) |
| Flash Point | >110°C (230°F) |
| Assay Percent Range | 0.96 |
| Refractive Index | 1.469 |
| Quantity | 25 g |
| Beilstein | 1753231 |
| Solubility Information | Soluble in water,0.2422mg/L @ 25°C (est). |
| Formula Weight | 233.2 |
| Percent Purity | 96% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction