Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11-Maleimidoundecanoic acid

Catalog No. p-7080191
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 57079-01-3 | C15H23NO4 | 281.352 g/mol

The maleimide functional group can be used to conjugate a variety of biomolecules such as enzymes and DNA to the polymer chain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57079-01-3
Molecular Formula C15H23NO4
Molecular Weight (g/mol) 281.352
MDL Number MFCD00941279
InChI Key UVZTZBRGZXIBLZ-UHFFFAOYSA-N
Synonym 11-maleimidoundecanoic acid, 11-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl undecanoic acid, unii-lg78ey7hvb, n-10-carboxydecyl maleimide, 11-2,5-dioxopyrrol-1-yl undecanoic acid, lg78ey7hvb, maleimide-ch2 10-cooh, 1h-pyrrole-1-undecanoic acid, 2,5-dihydro-2,5-dioxo, kmua, pubchem11837
PubChem CID 4618600
IUPAC Name 11-(2,5-dioxopyrrol-1-yl)undecanoic acid
SMILES C1=CC(=O)N(C1=O)CCCCCCCCCCC(=O)O

Specifications

Melting Point 89°C to 90°C
Color White
Quantity 5 g
Solubility Information Soluble in methanol,and chloroform.
Formula Weight 281.35
Physical Form Powder
Chemical Name or Material 11-Maleimidoundecanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.