Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

12-Crown-4, 97%, ACROS Organics™

Click to view available options
Quantity:
2.5g
5g
25g
Packaging:
Glass bottle
294-93-9
C8H16O4
176.21
MFCD00005103
XQQZRZQVBFHBHL-UHFFFAOYSA-N
12-crown-4,eoct,12-crown-4 ether,ethylene oxide cyclic tetramer,ccris 157,unii-k6069p2c2a,12-crown4-ether,12-crown 4-ether,5-19-11-00334 beilstein handbook reference,3,6,9,12-tetraoxacyclododecan
9269
CHEBI:32399
1,4,7,10-tetraoxacyclododecane
C1COCCOCCOCCO1
294-93-9
C8H16O4
176.21
MFCD00005103
XQQZRZQVBFHBHL-UHFFFAOYSA-N
12-crown-4,eoct,12-crown-4 ether,ethylene oxide cyclic tetramer,ccris 157,unii-k6069p2c2a,12-crown4-ether,12-crown 4-ether,5-19-11-00334 beilstein handbook reference,3,6,9,12-tetraoxacyclododecan
9269
CHEBI:32399
1,4,7,10-tetraoxacyclododecane
C1COCCOCCOCCO1
Chelation/Complexation reagent
Flash Point >112°C
Melting Point 16°C
Refractive Index 1.4600 to 1.463
Boiling Point 238°C
Density 1.0800g/mL
Specific Gravity 1.08
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 2.5g
Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Fieser 06,133; 09,126; 12,147; 13,167
Solubility Information Solubility in water: miscible
Formula Weight 176.21
Physical Form Liquid After Melting
Percent Purity 97%
Chemical Name or Material 12-Crown-4
Show More Show Less

GHS H Statement:
Fatal if inhaled.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-036-5