Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

12-Lipoxygenase Antibody (OTI1C3), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP27016A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27016A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27016A30 Supplier Novus Biologicals Supplier No. NBP270165AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

12-Lipoxygenase Monoclonal specifically detects 12-Lipoxygenase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen 12-Lipoxygenase
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1C3
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 12(S)-lipoxygenase, 12-LOX, 12S-lipoxygenase, 12S-LOXarachidonate 12-lipoxygenase, 12S-type, arachidonate 12-lipoxygenase, EC 1.13.11, EC 1.13.11.31, LOG12, platelet 12-LOX, platelet-type 12-lipoxygenase, Platelet-type lipoxygenase 12
Gene Symbols ALOX12
Host Species Mouse
Immunogen Full length recombinant protein of human 12-Lipoxygenase (NP_000688) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 239
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.