Learn More
Description
Specifications
Specifications
| Antigen | 12S-HETE |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Whole rabbit serum |
| Gene Alias | (5Z,8Z,10E,12S,14Z)-12-hydroxyicosa-5,8,10,14-tetraenoic acid, 12(S)-HETE, 12(S)-Hydroxyeicosatetraenoic acid, 12-Hete, 12-Hydroxyeicosatetraenoic acid, 12S-HETE, 54397-83-0, CHEBI:34146 |
| Host Species | Rabbit |
| Immunogen | 12S-HETE conjugated to human serum albumin. |
| Purification Method | Unpurified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
