Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GVS Poretics™ 13 mm Polycarbonate Disc
Shop All Osmonics Products
Click to view available options
Pore Size:
0.1 μm
0.4 μm
1.0 μm
2 μm
5 μm
8 μm
Description
Properties
- The polycarbonate membrane is manufactured by GVS using a two-stage production process that applies high quality standards.
- This unique process allows greater control over pore size and density to ensure that the physical properties of each membrane precisely match your specifications.
- The resulting membrane is a thin, translucent, microporous polycarbonate film with a smooth, flat surface. All particles larger than pore size are captured on its surface. It is ideal for use when collecting samples for blood tests or for high purity, general purpose filtration.
Characteristics
- Absolute pore size and density provides flow control for liquids moving through the membrane capturing 100 percent of cells larger than pore size.
- Smooth, thin, glass-like surface planar surface makes it ideal for particle identification by microscopy.
- Superior strength tensile strength of 207 bar (> 3000 psi) maintains pore size and density, and will not stretch.
- Low extractables ensure tests will be clean promoting consistent results.
- Negligible absorption/adsorption maximizes critical solution recovery.
- Available as hydrophilic or hydrophobic allows for a wide range of product applications.
Applications
- Flow control of reagents through assay
- General filtration
- Precise filtration and prefiltration
- Removal of red blood cells from plasma
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Disposable | Limited-use |
| Filter Type | Membrane Filter |
| Format | Disc |
| Material | PCTE |
| Pore Size | 1.0 μm |
| Sterility | Non-sterile |
| Type | Disc Membrane |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction