Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB062153 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB062153 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB062153 Supplier Novus Biologicals Supplier No. NBP299637C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

14-3-3 Polyclonal specifically detects 14-3-3 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen 14-3-3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias 14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant 14-3-3 (P27348; Met1-Asn245).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10971
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.