Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 beta/alpha Antibody (6F6), Novus Biologicals™
SDP

Catalog No. NB298483 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298483 100 μL
NB298482 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298483 Supplier Novus Biologicals Supplier No. NBP326247100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

14-3-3 beta/alpha Monoclonal antibody specifically detects 14-3-3 beta/alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry

Specifications

Antigen 14-3-3 beta/alpha
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 6F6
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias 14-3-3 alpha, 14-3-3 beta, 14-3-3 protein beta/alpha, brain protein 14-3-3, beta isoform, GW128, HS1, KCIP-1, Protein 1054, Protein kinase C inhibitor protein 1, protein kinase C inhibitor protein-1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, alphapolypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, betapolypeptide, YWHAA
Host Species Rabbit
Immunogen A synthesized peptide derived from Human 14-3-3 beta/alpha [UniProt P31946]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7529
Target Species Human, Mouse, Rat
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.