Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 beta/alpha Antibody (6F6), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | 14-3-3 beta/alpha |
---|---|
Clone | 6F6 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
14-3-3 beta/alpha Monoclonal antibody specifically detects 14-3-3 beta/alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
14-3-3 beta/alpha | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
14-3-3 alpha, 14-3-3 beta, 14-3-3 protein beta/alpha, brain protein 14-3-3, beta isoform, GW128, HS1, KCIP-1, Protein 1054, Protein kinase C inhibitor protein 1, protein kinase C inhibitor protein-1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, alphapolypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, betapolypeptide, YWHAA | |
A synthesized peptide derived from Human 14-3-3 beta/alpha [UniProt P31946] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
6F6 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
7529 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title