Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 beta/epsilon/zeta, Mouse anti-Human,Mouse,Rat, Biotin, Clone: 3C8, Novus Biologicals™
SDP

Catalog No. NB004741 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB004741 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB004741 Supplier Novus Biologicals Supplier No. NBP197885B

Mouse Monoclonal Antibody

14-3-3 beta/epsilon/zeta Monoclonal specifically detects 14-3-3 beta/epsilon/zeta in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen 14-3-3 beta/epsilon/zeta
Applications Western Blot
Classification Monoclonal
Clone 3C8
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Immunogen Recombinant human 14-3-3 beta.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.