Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 epsilon Antibody (02), FITC, Novus Biologicals™
SDP

Catalog No. NB293443 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293443 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293443 Supplier Novus Biologicals Supplier No. NBP326078F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

14-3-3 epsilon Monoclonal antibody specifically detects 14-3-3 epsilon in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen 14-3-3 epsilon
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 02
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias 14-3-3 epsilon, 14-3-3 protein epsilon, FLJ45465, FLJ53559,14-3-3E, KCIP-1, MDCR, MDS, mitochondrial import stimulation factor L subunit, protein kinase C inhibitor protein-1, tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilonpolypeptide
Host Species Mouse
Immunogen Hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human 14-3-3 epsilon (Accession#: NP_006752.1, Met1-Gln255)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7531
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.