Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 epsilon Antibody (CPTC-YWHAE-1), Biotin, Novus Biologicals™
SDP

Catalog No. NB013122 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013122 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013122 Supplier Novus Biologicals Supplier No. NBP279944B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

14-3-3 epsilon Monoclonal specifically detects 14-3-3 epsilon in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen 14-3-3 epsilon
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CPTC-YWHAE-1
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 14-3-3 epsilon, 14-3-3 protein epsilon, FLJ45465, FLJ53559,14-3-3E, KCIP-1, MDCR, MDS, mitochondrial import stimulation factor L subunit, protein kinase C inhibitor protein-1, tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilonpolypeptide
Gene Symbols YWHAE
Host Species Mouse
Immunogen Recombinant human full-length 14-3-3 epsilon protein (Uniprot: P62268)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7531
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.