Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 eta Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB324242 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB324242 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB324242 Supplier Novus Biologicals Supplier No. NBP299642JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

14-3-3 eta Polyclonal antibody specifically detects 14-3-3 eta in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen 14-3-3 eta
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias 14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA114-3-3 eta
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human 14-3-3 eta (Uniprot#: Q04917; Gly2-Asn246).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7533
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.