Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 gamma, ac Val2 Antibody (KC21), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB003501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003501 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003501 Supplier Novus Biologicals Supplier No. NB100406AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

14-3-3 gamma (ac Val2) Monoclonal specifically detects 14-3-3 gamma (ac Val2) in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen 14-3-3 gamma (ac Val2)
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone KC21
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 14-3-3 gamma, 14-3-3 protein gamma, 14-3-3GAMMA, KCIP-1, Protein kinase C inhibitor protein 1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gammapolypeptide
Gene Symbols YWHAG
Host Species Mouse
Immunogen N-terminal fragment of human 14-3-3 gamma where the N-terminal Val was acetylated [UniProt# P61981]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Test Specificity This antibody is specific for the naturally occurring human form of 14-3-3 gamma, where the N-terminal Met is removed, resulting in an acetylated N-terminal Val. Unprocessed (non-modified) 14-3-3 gamma is not recognized by this antibody.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.