Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 gamma Antibody (HS23), Novus Biologicals™
SDP

Catalog No. NB100407SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100407SS 0.025 mL
NB100407 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100407SS Supplier Novus Biologicals Supplier No. NB100407SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 8 publications

14-3-3 gamma Monoclonal specifically detects 14-3-3 gamma in Bovine, Chicken, Human, Mouse, Rat, Yeast, Zebrafish samples. It is validated for Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen 14-3-3 gamma
Applications Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HS23
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:50 - 1:250, Immunoblotting
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. P61981
Gene Alias 14-3-3 gamma, 14-3-3 protein gamma, 14-3-3GAMMA, KCIP-1, Protein kinase C inhibitor protein 1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gammapolypeptide
Gene Symbols YWHAG
Host Species Mouse
Immunogen N-terminal fragment of human 14-3-3 gamma [Uniprot: P61981]
Molecular Weight of Antigen 33 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7532
Test Specificity NB100-407 reacts with the human form of intact, non-modified N-terminus 14-3-3 gamma. The naturally occurring form of 14-3-3 gamma in which the N-terminus Met is removed and the Val is acetylated is not recognized by this antibody.
Target Species Bovine, Mouse, Chicken, Human, Mouse, Zebrafish, Rat, Yeast, Zebrafish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.