Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 gamma Antibody (HS23), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 8 publications
$243.50 - $499.50
Specifications
Antigen | 14-3-3 gamma |
---|---|
Clone | HS23 |
Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:50 - 1:250, Immunoblotting |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100407SS
![]() |
Novus Biologicals
NB100407SS |
0.025 mL |
Each for $243.50
|
|
|||||
NB100407
![]() |
Novus Biologicals
NB100407 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
14-3-3 gamma Monoclonal specifically detects 14-3-3 gamma in Human, Mouse, Rat, Bovine, Chicken, Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoblotting.Specifications
14-3-3 gamma | |
Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:50 - 1:250, Immunoblotting | |
Monoclonal | |
Ascites | |
RUO | |
Human, Mouse, Rat, Bovine, Chicken, Zebrafish | |
P61981 | |
7532 | |
N-terminal fragment of human 14-3-3 gamma [Uniprot: P61981] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
33 kDa |
HS23 | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot | |
Unconjugated | |
Mouse | |
Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
Ascites with 0.1% Sodium Azide | |
14-3-3 gamma, 14-3-3 protein gamma, 14-3-3GAMMA, KCIP-1, Protein kinase C inhibitor protein 1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gammapolypeptide | |
YWHAG | |
IgG1 κ | |
Unpurified | |
NB100-407 reacts with the human form of intact, non-modified N-terminus 14-3-3 gamma. The naturally occurring form of 14-3-3 gamma in which the N-terminus Met is removed and the Val is acetylated is not recognized by this antibody. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title