Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 Rabbit anti-Human, Clone: JJ084-3, Novus Biologicals™
SDP

Catalog No. NBP267866 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267866 Supplier Novus Biologicals Supplier No. NBP267866

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

14-3-3 Monoclonal specifically detects 14-3-3 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen 14-3-3
Applications Western Blot
Classification Monoclonal
Clone JJ084-3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias 14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide
Gene Symbols YWHAQ
Host Species Rabbit
Immunogen Recombinant protein within Human 14-3-3 aa 1-122/ 246. (SwissProt: P31946 Human; SwissProt: P63104 Human; SwissProt: P63101 Mouse; SwissProt: Q9CQV8 Mouse; SwissProt: P35213 Rat; SwissProt: P63102 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10971
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.