Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 sigma/Stratifin Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB324301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB324301 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB324301 Supplier Novus Biologicals Supplier No. NBP299630MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

14-3-3 sigma/Stratifin Polyclonal antibody specifically detects 14-3-3 sigma/Stratifin in Human samples. It is validated for ELISA

Specifications

Antigen 14-3-3 sigma/Stratifin
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias 14-3-3 protein sigma, 14-3-3 sigma, Epithelial cell marker protein 1, HME1, stratifin, YWHAS
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human 14-3-3 sigma/Stratifin (Accession#: NP_006133.1; Met 1-Ser 248).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2810
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.