Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 sigma/Stratifin Antibody, Novus Biologicals™
SDP

Catalog No. NBP227072SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227072SS 0.025 mg
NBP227072 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227072SS Supplier Novus Biologicals Supplier No. NBP227072SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

14-3-3 sigma/Stratifin Polyclonal specifically detects 14-3-3 sigma/Stratifin in Human, Amphibian, Bovine, Canine, Equine, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen 14-3-3 sigma/Stratifin
Applications Western Blot, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-5ug/ml∼, Immunohistochemistry-Paraffin 5-10ug/ml, Knockdown Validated
Formulation PBS (pH 7.4) with 0.05% Sodium Azide
Gene Accession No. P31947
Gene Alias 14-3-3 protein sigma, 14-3-3 sigma, Epithelial cell marker protein 1, HME1, stratifin, YWHAS
Gene Symbols SFN
Host Species Rabbit
Immunogen A portion of amino acids 120-170 of human 14-3-3 sigma was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2810
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, mouse, rat, pig, new world monkey, panda, cow, dog, orangutan, rhesus monkey, white-cheeked gibbon, chimpanzee, rabbit, and sheep, 94% in horse, 89% in opossum, and 72% in lizard.This antibody will detect both isoforms of 14-3-3 sigma (248 amino acids and 169 amino acids).
Target Species Human, Amphibian, Bovine, Canine, Equine, Opossum, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.