Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 sigma/Stratifin Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen 14-3-3 sigma/Stratifin
Concentration 1.0 mg/mL
Dilution Western Blot 3-5ug/ml∼, Immunohistochemistry-Paraffin 5-10ug/ml, Knockdown Validated
Applications Western Blot, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227072SS
SDP
View Documents
Novus Biologicals
NBP227072SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP227072
SDP
View Documents
Novus Biologicals
NBP227072
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

14-3-3 sigma/Stratifin Polyclonal specifically detects 14-3-3 sigma/Stratifin in Human, Amphibian, Bovine, Canine, Equine, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

14-3-3 sigma/Stratifin
Western Blot 3-5ug/ml∼, Immunohistochemistry-Paraffin 5-10ug/ml, Knockdown Validated
Polyclonal
Purified
RUO
Human, Amphibian, Bovine, Canine, Equine, Opossum, Primate
P31947
2810
A portion of amino acids 120-170 of human 14-3-3 sigma was used as the immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/mL
Western Blot, Immunohistochemistry (Paraffin), KnockDown
Unconjugated
Rabbit
Cell Cycle and Replication, Phospho Specific
PBS (pH 7.4) with 0.05% Sodium Azide
14-3-3 protein sigma, 14-3-3 sigma, Epithelial cell marker protein 1, HME1, stratifin, YWHAS
SFN
IgG
Protein A purified
The amino acid sequence used as immunogen for the antibody is 100% homologous in human, mouse, rat, pig, new world monkey, panda, cow, dog, orangutan, rhesus monkey, white-cheeked gibbon, chimpanzee, rabbit, and sheep, 94% in horse, 89% in opossum, and 72% in lizard.This antibody will detect both isoforms of 14-3-3 sigma (248 amino acids and 169 amino acids).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.