Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 tau/theta Antibody (1A1), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB310159 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310159 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310159 Supplier Novus Biologicals Supplier No. NBP121301MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

14-3-3 tau/theta Monoclonal antibody specifically detects 14-3-3 tau/theta in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen 14-3-3 tau/theta
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1A1
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide
Host Species Mouse
Immunogen Recombinant human 14-3-3 tau/theta (1-245aa) purified from E. coli (NP_006817).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 10971
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.