Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 zeta Antibody, Novus Biologicals™
SDP

Catalog No. NBP224593SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224593SS 0.025 mg
NBP224593 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224593SS Supplier Novus Biologicals Supplier No. NBP224593SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

14-3-3 zeta Polyclonal specifically detects 14-3-3 zeta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen 14-3-3 zeta
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias KCIP-1MGC126532,14-3-3-zeta, MGC111427, MGC138156, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, deltapolypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zetapolypeptide, zeta polypeptide
Gene Symbols YWHAZ
Host Species Rabbit
Immunogen A portion of amino acids 200-245 of human 14-3-3 zeta was used as the immunogen for the antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 7534
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human and 94% homologous in mouse, rat and bovine.
Target Species Human, Mouse, Rat, Bovine, Canine, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.