Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3beta/alpha Mouse anti-Human, Clone: 43369, Novus Biologicals™
SDP

Catalog No. NBP267090 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267090 Supplier Novus Biologicals Supplier No. NBP267090

Mouse Monoclonal Antibody

14-3-3 beta/alpha Monoclonal specifically detects 14-3-3 beta/alpha in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen 14-3-3beta/alpha
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 43369
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200
Formulation PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias 14-3-3 alpha, 14-3-3 beta, 14-3-3 protein beta/alpha, brain protein 14-3-3, beta isoform, GW128, HS1, KCIP-1, Protein 1054, Protein kinase C inhibitor protein 1, protein kinase C inhibitor protein-1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, alphapolypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, betapolypeptide, YWHAA
Gene Symbols YWHAB
Host Species Mouse
Immunogen Recombinant protein within Human 14-3-3 beta/alpha aa 1-246/ 246. (SwissProt: P31946 Human)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7529
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.